Tisk…

3-(Morpholinosulphonyl)phenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: (3-(Morpholinosulphonyl)phenyl)boronic acid, 3-(4-Morpholinylsulphonyl)benzeneboronic acid

OR5713-250MG
APOSOR5713-250MGEA 1820 CZK
APOSOR5713-250MG
3-(Morpholinosulphonyl)phenylboronic acid 98%
3-(Morpholinosulphonyl)phenylboronic acid
Vzorec: C₁₀H₁₄BNO₅S
MW: 271,09 g/mol
Bod tání: 144…148 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363748
CAS číslo: 871329-60-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR