Tisk…

4-Carboxy-2-chlorophenylboronic acid 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-Carboxy-2-chlorobenzeneboronic acid

OR5712-25G OR5712-5G OR5712-100G
APOSOR5712-25GEA 6290 CZK
APOSOR5712-25G APOSOR5712-5G APOSOR5712-100G
4-Carboxy-2-chlorophenylboronic acid 98%
4-Carboxy-2-chlorophenylboronic acid
Vzorec: C₇H₆BClO₄
MW: 200,39 g/mol
Bod tání: 208…210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363763
CAS číslo: 851335-09-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR