Tisk…

4-Bromo-N-(2-(TBDMSO)ethyl)benzenesulphonamide 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Bromo-N-[2-(tert-butyldimethylsilyloxy)ethyl]benzenesulphonamide, 4-Bromo-N-(2-((tert-butyldimethylsilyl)oxy)ethyl)benzenesulphonamide

OR5709-100G OR5709-25G OR5709-5G
APOSOR5709-100GEA 10650 CZK
APOSOR5709-100G APOSOR5709-25G APOSOR5709-5G
4-Bromo-N-(2-(TBDMSO)ethyl)benzenesulphonamide 98%
4-Bromo-N-(2-(TBDMSO)ethyl)benzenesulphonamide
Vzorec: C₁₄H₂₄BrNO₃SSi
MW: 394,400 g/mol
Bod tání: 32…36 °C
Storage Temperature: Ambient
MDL Number: MFCD06659896
CAS číslo: 850429-52-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR