N-Benzyl-4-bromobenzenesulphonamide 98%
Dodavatel: Apollo Scientific
OR5707-25G
OR5707-5G
APOSOR5707-25GEA
7510
CZK
APOSOR5707-25G
APOSOR5707-5G
N-Benzyl-4-bromobenzenesulphonamide 98%
N-Benzyl-4-bromobenzenesulphonamide
Vzorec:
C₁₃H₁₂BrNO₂S MW: 326,21 g/mol Bod tání: 102…104 °C Storage Temperature: Ambient |
MDL Number:
MFCD00450587 CAS číslo: 3609-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...