4-Bromo-N-butylbenzenesulphonamide 98%
Dodavatel: Apollo Scientific
OR5701-25G
OR5701-5G
APOSOR5701-25GEA
14920
CZK
APOSOR5701-25G
APOSOR5701-5G
4-Bromo-N-butylbenzenesulphonamide 98%
4-Bromo-N-butylbenzenesulphonamide
Vzorec:
C₁₀H₁₄BrNO₂S Bod tání: 51…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD01215086 CAS číslo: 1984-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...