Tisk…

4-Bromo-N,N-dimethylbenzenesulphonamide 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: N,N-Dimethyl 4-bromobenzenesulphonamide

OR5693-1G OR5693-5G
APOSOR5693-1GEA 622 CZK
APOSOR5693-1G APOSOR5693-5G
4-Bromo-N,N-dimethylbenzenesulphonamide 97%
4-Bromo-N,N-dimethylbenzenesulphonamide
Vzorec: C₈H₁₀BrNO₂S
MW: 264,14 g/mol
Bod tání: 88…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00017834
CAS číslo: 707-60-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR