1-Benzylpiperidine-4-carboxylic acid 96%
Dodavatel: Apollo Scientific
|
Varování
|
OR5690-1G
OR5690-5G
APOSOR5690-1GEA
622
CZK
APOSOR5690-1G
APOSOR5690-5G
1-Benzylpiperidine-4-carboxylic acid 96%
1-Benzylpiperidine-4-carboxylic acid
Vzorec:
C₁₃H₁₇NO₂ MW: 219,28 g/mol Bod tání: 167…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD03371463 CAS číslo: 10315-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...