Tisk…

1-(Pyridin-2-ylmethyl)piperidine-4-carboxylic acid 95+%

Dodavatel: Apollo Scientific

OR5688-10G OR5688-1G OR5688-5G
APOSOR5688-10GEA 13960 CZK
APOSOR5688-10G APOSOR5688-1G APOSOR5688-5G
1-(Pyridin-2-ylmethyl)piperidine-4-carboxylic acid 95+%
1-(Pyridin-2-ylmethyl)piperidine-4-carboxylic acid
Vzorec: C₁₂H₁₆N₂O₂
MW: 220,2700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03371469
CAS číslo: 193538-28-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR