3-Methyl-6-nitrocoumarin 95+%
Dodavatel: Apollo Scientific
OR5669-1G
OR5669-5G
OR5669-25G
APOSOR5669-1GEA
3870
CZK
APOSOR5669-1G
APOSOR5669-5G
APOSOR5669-25G
3-Methyl-6-nitrocoumarin 95+%
3-Methyl-6-nitrocoumarin
Vzorec:
C₁₀H₇NO₄ Bod tání: 220…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD01941323 CAS číslo: 95532-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...