Tisk…

[(1-Methyl-1H-indol-2-yl)methyl]amine 96%

Dodavatel: Apollo Scientific
Varování

Synonyms: (1-Methyl-1H-indol-2-yl)methanamine, 2-(Aminomethyl)-1-methyl-1H-indole

OR5663-250MG OR5663-100MG OR5663-1G
APOSOR5663-250MGEA 2940 CZK
APOSOR5663-250MG APOSOR5663-100MG APOSOR5663-1G
[(1-Methyl-1H-indol-2-yl)methyl]amine 96%
[(1-Methyl-1H-indol-2-yl)methyl]amine
Vzorec: C₁₀H₁₂N₂
MW: 160,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06801045
CAS číslo: 55556-57-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR