Tisk…

Piperidin-1-ylacetic acid hydrochloride

Dodavatel: Apollo Scientific
Varování

OR5652-1G OR5652-250MG
APOSOR5652-1GEA 827 CZK
APOSOR5652-1G APOSOR5652-250MG
Piperidin-1-ylacetic acid hydrochloride
Piperidin-1-ylacetic acid hydrochloride
Vzorec: C₇H₁₄ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD01701566
CAS číslo: 3235-68-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR