Tisk…

Cyclohexylhydrazine hydrochloride

Dodavatel: Apollo Scientific
Nebezpečí

OR5641-1G OR5641-25G
APOSOR5641-1GEA 622 CZK
APOSOR5641-1G APOSOR5641-25G
Cyclohexylhydrazine hydrochloride
Cyclohexylhydrazine hydrochloride
Vzorec: C₆H₁₄N₂
MW: 150.65 g/mol
Bod tání: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00060160
CAS číslo: 24214-73-1
UN: 3263
ADR: 8 II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR