Tisk…

2,3-Dimethyl-5-nitroindole

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,3-Dimethyl-5-nitro-1H-indole

OR5638-250MG OR5638-5G
APOSOR5638-250MGEA 0 CZK
APOSOR5638-250MG APOSOR5638-5G
2,3-Dimethyl-5-nitroindole
2,3-Dimethyl-5-nitroindole
Vzorec: C₁₀H₁₀N₂O₂
Bod tání: 184…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00022708
CAS číslo: 21296-94-6
EINECS: 244-321-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR