trans-4-Aminocyclohexanol 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR5631-25G
OR5631-100G
OR5631-500G
APOSOR5631-25GEA
4770
CZK
APOSOR5631-25G
APOSOR5631-100G
APOSOR5631-500G
trans-4-Aminocyclohexanol 97%
trans-4-Aminocyclohexanol
Vzorec:
C₆H₁₃NO Bod varu: 124 °C/14mmHg Bod tání: 108…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067698 CAS číslo: 27489-62-9 EINECS: 249-492-8 UN: 3259 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...