Methyl-2-boc-aminothiazole-4-carboxylate 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Methyl 2-Boc-aminothiazole-4-carboxylate, Methyl 2-amino-1,3-thiazole-4-carboxylate, 2-BOC protected
OR5619-1G
OR5619-5G
APOSOR5619-1GEA
1910
CZK
APOSOR5619-1G
APOSOR5619-5G
Methyl-2-boc-aminothiazole-4-carboxylate 98%
Methyl-2-boc-aminothiazole-4-carboxylate
Vzorec:
C₁₀H₁₄N₂O₄S MW: 258,2942 g/mol Bod tání: 156…160 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659910 CAS číslo: 850429-62-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...