4-(Hydroxymethyl)thiazole 95%
Dodavatel: Apollo Scientific
|
Varování
|
OR5598-1G
OR5598-25G
OR5598-5G
APOSOR5598-1GEA
4170
CZK
APOSOR5598-1G
APOSOR5598-25G
APOSOR5598-5G
4-(Hydroxymethyl)thiazole 95%
4-(Hydroxymethyl)thiazole
Vzorec:
C₄H₅NOS MW: 115,15 g/mol Bod varu: 85…90 °C/0,5mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD04115728 CAS číslo: 7036-04-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...