Tisk…

5-Thiazolemethanol 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Thiazol-5-ylmethanol, 5-(Hydroxymethyl)thiazole, Thiazole-5-methanol

OR5597-100G OR5597-25G OR5597-5G
APOSOR5597-100GEA 2280 CZK
APOSOR5597-100G APOSOR5597-25G APOSOR5597-5G
5-Thiazolemethanol 98%
5-Thiazolemethanol
Vzorec: C₄H₅NOS
MW: 115,15 g/mol
Bod varu: 106 °C/0,075mmHg
Hustota: 1,32 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD04115732
CAS číslo: 38585-74-9
EINECS: 414-780-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR