3-(4-sulphamoylphenyl)propionic acid 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
3-(4-sulphamoylphenyl)propanoic acid
OR5594-1G
OR5594-5G
APOSOR5594-1GEA
1050
CZK
APOSOR5594-1G
APOSOR5594-5G
3-(4-sulphamoylphenyl)propionic acid 98%
3-(4-sulphamoylphenyl)propionic acid
Vzorec:
C₉H₁₁NO₄S MW: 229.26 g/mol Bod tání: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659895 CAS číslo: 90610-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...