Tisk…

Ethyl-p-tolylacetate 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Ethyl p-Tolylacetate, (4-Methylphenyl)acetic acid ethyl ester, p-Tolylacetic acid ethyl ester

OR5589-10G OR5589-50G OR5589-5G OR5589-25G
APOSOR5589-10GEA 1290 CZK
APOSOR5589-10G APOSOR5589-50G APOSOR5589-5G APOSOR5589-25G
Ethyl-p-tolylacetate 98%
Ethyl-p-tolylacetate
Vzorec: (C₆H₄(CH₃))CH₂COOCH₂CH₃
MW: 178,23 g/mol
Bod varu: 240…241 °C
Hustota: 1,008 g/cm³ (25 °C)
Bod vzplanutí: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00009181
CAS číslo: 14062-19-2
EINECS: 237-904-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR