5-Thiazolecarboxylic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR5586-5G
OR5586-1G
OR5586-25G
APOSOR5586-5GEA
2150
CZK
APOSOR5586-5G
APOSOR5586-1G
APOSOR5586-25G
5-Thiazolecarboxylic acid 97%
5-Thiazolecarboxylic acid
Vzorec:
C₄H₃NO₂S MW: 129,1371 g/mol Bod tání: 190 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03428539 CAS číslo: 14527-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...