Tisk…

Ethyl-2-aminoisonicotinate 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 2-Amino-isonicotinic acid ethyl ester, Ethyl 2-aminopyridine-4-carboxylate, 2-Aminopyridine-4-carboxylic acid Ethyl Ester, Ethyl 2-aminoisonicotinate, Ethyl 2-Aminopyridine-4-carboxylate, 2-Aminoisonicotinic acid Ethyl Ester, Ethyl 2-Aminoisonicotinate

OR5579-1G OR5579-10G OR5579-5G
APOSOR5579-1GEA 622 CZK
APOSOR5579-1G APOSOR5579-10G APOSOR5579-5G
Ethyl-2-aminoisonicotinate 97%
Ethyl 2-aminoisonicotinate
Vzorec: C₈H₁₀N₂O₂
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03791260
CAS číslo: 13362-30-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR