2-Acetylamino-4-methylpyridine 98%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
N-(4-Methyl-2-pyridyl)acetamide, N-(4-Methylpyridin-2-yl)acetamide, 2-Acetamido-4-picoline, 2-Acetamido-4-methylpyridine
OR5569-100G
OR5569-25G
OR5569-5G
APOSOR5569-100GEA
22520
CZK
APOSOR5569-100G
APOSOR5569-25G
APOSOR5569-5G
2-Acetylamino-4-methylpyridine 98%
2-Acetylamino-4-methylpyridine
Vzorec:
C₈H₁₀N₂O MW: 150,1778 g/mol Bod tání: 90…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234432 CAS číslo: 5327-32-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...