4-Bromopicolinic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Bromopyridine-2-carboxylic acid
OR5568-10G
OR5568-25G
OR5568-5G
APOSOR5568-10GEA
910
CZK
APOSOR5568-10G
APOSOR5568-25G
APOSOR5568-5G
4-Bromopicolinic acid 97%
4-Bromopicolinic acid
Vzorec:
C₆H₄BrNO₂ MW: 202,01 g/mol Bod tání: 172…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426665 CAS číslo: 30766-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...