Tisk…

Sodium-4-chlorobenzenesulphonate 98%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Chlorobenzenesulphonic acid Sodium Salt

OR55632-25G OR55632-500G
APOSOR55632-25GEA 1450 CZK
APOSOR55632-25G APOSOR55632-500G
Sodium-4-chlorobenzenesulphonate 98%
Sodium-4-chlorobenzenesulphonate
Vzorec: C₆H₅ClO₃S
MW: 214.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014731
CAS číslo: 5138-90-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR