4-Bromo-5-chloro-1H-indazole
Dodavatel: Apollo Scientific
|
Varování
|
OR55606-1G
OR55606-250MG
APOSOR55606-1GEA
6520
CZK
APOSOR55606-1G
APOSOR55606-250MG
4-Bromo-5-chloro-1H-indazole
4-Bromo-5-chloro-1H-indazole
Vzorec:
C₇H₄BrClN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD12828060 CAS číslo: 1056264-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...