Tisk…

5-Bromo-4-chloro-1H-indazole

Dodavatel: Apollo Scientific
Varování

OR55605-1G OR55605-250MG OR55605-5G
APOSOR55605-1GEA 1410 CZK
APOSOR55605-1G APOSOR55605-250MG APOSOR55605-5G
5-Bromo-4-chloro-1H-indazole
5-Bromo-4-chloro-1H-indazole
Vzorec: C₇H₄BrClN₂
MW: 231,4771 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11007933
CAS číslo: 1082041-90-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR