Tisk…

Methyl-2-bromo-4-methoxybenzoate

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2-Bromo-4-methoxybenzoic acid methyl ester, Methyl 2-bromo-4-methoxybenzoate

OR55591-1G OR55591-5G OR55591-10G
APOSOR55591-1GEA 705 CZK
APOSOR55591-1G APOSOR55591-5G APOSOR55591-10G
Methyl-2-bromo-4-methoxybenzoate
Methyl-2-bromo-4-methoxybenzoate
Vzorec: C₉H₉BrO₃
MW: 245,07 g/mol
Bod tání: 25…26 °C
Storage Temperature: Ambient
MDL Number: MFCD12026001
CAS číslo: 17100-65-1
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR