Tisk…

(3-Nitrophenyl)methanesulphonyl chloride 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 3-Nitro-α-toluenesulphonyl chloride

OR55548-25G OR55548-5G
APOSOR55548-25GEA 20500 CZK
APOSOR55548-25G APOSOR55548-5G
(3-Nitrophenyl)methanesulphonyl chloride 95%
(3-Nitrophenyl)methanesulphonyl chloride
Vzorec: C₇H₆ClNO₄S
MW: 235,64 g/mol
Bod tání: 95…102 °C
Storage Temperature: Ambient
MDL Number: MFCD01859935
CAS číslo: 58032-84-1
UN: 3261
ADR: 8,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR