Tisk…

5-Bromo-4-methoxy-2-nitroaniline 98%

Dodavatel: Apollo Scientific

OR55519-1G OR55519-250MG
APOSOR55519-1GEA 13690 CZK
APOSOR55519-1G APOSOR55519-250MG
5-Bromo-4-methoxy-2-nitroaniline 98%
5-Bromo-4-methoxy-2-nitroaniline
Vzorec: C₇H₇BrN₂O₃
MW: 247,0461 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00031127
CAS číslo: 6943-69-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR