3-(Bromomethyl)-1-methylpiperidine hydrobromide 99%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR55482-1G
OR55482-250MG
OR55482-5G
APOSOR55482-1GEA
8070
CZK
APOSOR55482-1G
APOSOR55482-250MG
APOSOR55482-5G
3-(Bromomethyl)-1-methylpiperidine hydrobromide 99%
3-(Bromomethyl)-1-methylpiperidine hydrobromide
Vzorec:
C₇H₁₅Br₂N Storage Temperature: Ambient |
MDL Number:
MFCD00101673 CAS číslo: 41886-04-8 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...