5-Hydroxypicolinic acid 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
5-hydroxypyridine-2-carboxylic acid
OR5545-1G
OR5545-5G
OR5545-100G
OR5545-25G
APOSOR5545-1GEA
622
CZK
APOSOR5545-1G
APOSOR5545-5G
APOSOR5545-100G
APOSOR5545-25G
5-Hydroxypicolinic acid 95%
5-Hydroxypicolinic acid
Vzorec:
C₆H₅NO₃ Bod tání: 260…264 °C Storage Temperature: Ambient |
MDL Number:
MFCD00661309 CAS číslo: 15069-92-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...