5-(Boc-amino)-2-(hydroxymethyl)benzeneboronic acid hemiester 98%
Dodavatel: Apollo Scientific
Synonyms:
5-Amino-2-(hydroxymethyl)benzeneboronic acid, dehydrate, N-BOC protected
OR5544-1G
OR5544-250MG
APOSOR5544-1GEA
12300
CZK
APOSOR5544-1G
APOSOR5544-250MG
5-(Boc-amino)-2-(hydroxymethyl)benzeneboronic acid hemiester 98%
5-(Boc-amino)-2-(hydroxymethyl)benzeneboronic acid hemiester
Vzorec:
C₁₂H₁₆BNO₄ MW: 249,07 g/mol Bod tání: 134…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659853 CAS číslo: 850568-79-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...