2'-Aminoacetophenone 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(2-Aminophenyl)ethanone, o-Aminoacetophenone
OR55399-100G
OR55399-1KG
OR55399-500G
APOSOR55399-100GEA
1120
CZK
APOSOR55399-100G
APOSOR55399-1KG
APOSOR55399-500G
2'-Aminoacetophenone 95%
2'-Aminoacetophenone
Vzorec:
C₈H₉NO Storage Temperature: Refrigerator |
MDL Number:
MFCD00007717 CAS číslo: 551-93-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...