5-Acetamido-2-nitrophenylboronic acid 96%
Dodavatel: Apollo Scientific
Synonyms:
(5-Acetamido-2-nitro)benzeneboronicacid
OR5539-1G
OR5539-5G
OR5539-10G
APOSOR5539-1GEA
4070
CZK
APOSOR5539-1G
APOSOR5539-5G
APOSOR5539-10G
5-Acetamido-2-nitrophenylboronic acid 96%
5-Acetamido-2-nitrophenylboronic acid
Vzorec:
C₈H₉BN₂O₅ MW: 223.9800 g/mol Bod tání: 193…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659829 CAS číslo: 78887-36-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...