Tisk…

5,10,15,20-Tetra(4-pyridyl)porphyrin

Dodavatel: Apollo Scientific
Varování

Synonyms: 5,10,15,20-Tetra(4-pyridyl)-21H,23H-porphine, meso-Tetra(4-pyridyl)porphine

OR55340-100MG OR55340-250MG
APOSOR55340-100MGEA 622 CZK
APOSOR55340-100MG APOSOR55340-250MG
5,10,15,20-Tetra(4-pyridyl)porphyrin
5,10,15,20-Tetra(4-pyridyl)porphyrin
Vzorec: C₄₀H₂₆N₈
MW: 618,69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00009626
CAS číslo: 16834-13-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR