Tisk…

5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin

Dodavatel: Apollo Scientific
Varování

Synonyms: 5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphine

OR55339-100MG OR55339-250MG
APOSOR55339-100MGEA 5200 CZK
APOSOR55339-100MG APOSOR55339-250MG
5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin
5,10,15,20-Tetrakis(2,6-dichlorophenyl)porphyrin
Vzorec: C₄₄H₂₂Cl₈N₄
MW: 890,29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191500
CAS číslo: 37083-37-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR