Quinoline-N-oxide hydrate
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Quinoline 1-oxide hydrate
OR55323-25G
OR55323-5G
APOSOR55323-25GEA
8300
CZK
APOSOR55323-25G
APOSOR55323-5G
Quinoline-N-oxide hydrate
Quinoline-N-oxide hydrate
Vzorec:
C₉H₇NO·xH₂O MW: 145.16 g/mol (anhy) Bod tání: 52…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149472 CAS číslo: 64201-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...