Tisk…

1-(1-Ethoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

Synonyms: 1-(1-Ethoxyethyl)-1H-pyrazole-4-boronic acid pinacol ester

OR55282-100G
APOSOR55282-100GEA 3150 CZK
APOSOR55282-100G
1-(1-Ethoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
1-(1-Ethoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Vzorec: C₁₃H₂₃BN₂O₃
MW: 266,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11857753
CAS číslo: 1029716-44-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR