Tisk…

Isoxazolidine hydrochloride

Dodavatel: Apollo Scientific

OR55263-250MG OR55263-1G OR55263-5G
APOSOR55263-250MGEA 1750 CZK
APOSOR55263-250MG APOSOR55263-1G APOSOR55263-5G
Isoxazolidine hydrochloride
Isoxazolidine hydrochloride
Vzorec: C₃H₇NO·HCl
MW: 109.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11100465
CAS číslo: 39657-45-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR