Tisk…

(S)-Pyrrolidine-2-carbonitrile hydrochloride 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: (S)-2-Pyrrolidinecarbonitrile hydrochloride

OR55254-250MG OR55254-1G OR55254-5G
APOSOR55254-250MGEA 1700 CZK
APOSOR55254-250MG APOSOR55254-1G APOSOR55254-5G
(S)-Pyrrolidine-2-carbonitrile hydrochloride 95%
(S)-Pyrrolidine-2-carbonitrile hydrochloride
Vzorec: C₅H₉ClN₂
MW: 132,5914 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02683060
CAS číslo: 65732-69-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR