Tisk…

7-Octenoic acid

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: Oct-7-enoic acid

OR55219-1G OR55219-5G
APOSOR55219-1GEA 2750 CZK
APOSOR55219-1G APOSOR55219-5G
7-Octenoic acid
7-Octenoic acid
Vzorec: C₈H₁₄O₂
MW: 142.2 g/mol
Hustota: 0.925 g/cm³ (25 °C)
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02258724
CAS číslo: 18719-24-9
UN: 3265
ADR: 8 III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR