Tisk…

4-Chloro-1H-pyrrolo[2,3-c]pyridine

Dodavatel: Apollo Scientific

OR55213-1G OR55213-5G
APOSOR55213-1GEA 3060 CZK
APOSOR55213-1G APOSOR55213-5G
4-Chloro-1H-pyrrolo[2,3-c]pyridine
4-Chloro-1H-pyrrolo[2,3-c]pyridine
Vzorec: C₇H₅ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD16657181
CAS číslo: 1188313-15-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR