Octadecylphosphonic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
n-Octadecylphosphonic acid, ODPA
OR55172-5G
OR55172-25G
APOSOR55172-5GEA
622
CZK
APOSOR55172-5G
APOSOR55172-25G
Octadecylphosphonic acid 97%
Octadecylphosphonic acid
Vzorec:
C₁₈H₃₉O₃P MW: 334.47 g/mol Bod tání: 92…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015838 CAS číslo: 4724-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...