6-Bromo-2-chloroquinazoline 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR55165-10G
OR55165-1G
OR55165-5G
APOSOR55165-10GEA
4300
CZK
APOSOR55165-10G
APOSOR55165-1G
APOSOR55165-5G
6-Bromo-2-chloroquinazoline 97%
6-Bromo-2-chloroquinazoline
Vzorec:
C₈H₄BrClN₂ MW: 243,4900 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09261000 CAS číslo: 882672-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...