Tisk…

BAPTA tetramethyl ester 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: Tetramethyl 2,2',2'',2'''-(((ethane-1,2-diylbis(oxy))bis(2,1-phenylene))bis(azanetriyl))tetraacetate

OR55133-1G OR55133-5G
APOSOR55133-1GEA 1080 CZK
APOSOR55133-1G APOSOR55133-5G
BAPTA tetramethyl ester 98%
BAPTA tetramethyl ester
Vzorec: C₂₆H₃₂N₂O₁₀
MW: 532,546 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00532664
CAS číslo: 125367-34-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR