Tisk…

6-Bromo-1-methyl-1H-indazole-3-carboxylic acid

Dodavatel: Apollo Scientific
Varování

OR55104-100MG OR55104-250MG OR55104-1G
APOSOR55104-100MGEA 2980 CZK
APOSOR55104-100MG APOSOR55104-250MG APOSOR55104-1G
6-Bromo-1-methyl-1H-indazole-3-carboxylic acid
6-Bromo-1-methyl-1H-indazole-3-carboxylic acid
Vzorec: C₉H₇BrN₂O₂
MW: 255,08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15071436
CAS číslo: 1021859-29-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR