Tisk…

Tigloyl chloride

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: trans-2-Methyl-2-butenoyl Chloride

OR55081-1G OR55081-5G
APOSOR55081-1GEA 2790 CZK
APOSOR55081-1G APOSOR55081-5G
Tigloyl chloride
Tigloyl chloride
Vzorec: C₅H₇ClO
Bod varu: 145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00800725
CAS číslo: 35660-94-7
UN: 3286
ADR: 3+6.1+8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR