2-Chloromethyl-5-methylpyrimidine hydrochloride
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-(Chloromethyl)-5-methylpyrimidine hydrochloride
OR55033-1G
OR55033-250MG
APOSOR55033-1GEA
13690
CZK
APOSOR55033-1G
APOSOR55033-250MG
2-Chloromethyl-5-methylpyrimidine hydrochloride
2-Chloromethyl-5-methylpyrimidine hydrochloride
Vzorec:
C₆H₈Cl₂N₂ MW: 179,05 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD22196490 CAS číslo: 1384430-75-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...