Tisk…

(S)-2,3-Diaminopropanoic acid hydrochloride 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: H-Dap-OH.HCl, H-Dap-OH·HCl, L-3-Aminoalanine Hydrochloride, (S)-2,3-Diaminopropionic acid monohydrochloride

OR5495-100G OR5495-25G
APOSOR5495-100GEA 8370 CZK
APOSOR5495-100G APOSOR5495-25G
(S)-2,3-Diaminopropanoic acid hydrochloride 98%
(S)-2,3-Diaminopropanoic acid hydrochloride
Vzorec: C₃H₈N₂O₂·HCl
MW: 140,6 g/mol
Bod tání: 231…233 °C
Storage Temperature: Ambient
MDL Number: MFCD00065497
CAS číslo: 1482-97-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR