Furosemide
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
5-(Aminosulphonyl)-4-chloro-2-[(2-furanylmethyl)amino]benzoic acid, 4-Chloro-N-furfuryl-5-sulphamoylanthranilic acid
OR5475-100G
OR5475-25G
OR5475-5G
APOSOR5475-100GEA
5720
CZK
APOSOR5475-100G
APOSOR5475-25G
APOSOR5475-5G
Furosemide
Furosemide
Vzorec:
C₁₂H₁₁ClN₂O₅S MW: 330,74 g/mol Bod tání: 220 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00010549 CAS číslo: 54-31-9 EINECS: 200-203-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...